Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACCCCTCCAGGCTCACACTCTG[A/G]GCCCTGCGCCGACACTCAACTGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600566 MIM: 605577 | ||||||||||||||||||||
Literature Links: |
NRXN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NRXN2 - neurexin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASGRP2 - RAS guanyl releasing protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098670.1 | 2234 | Silent Mutation | GCC,GCT | A,A 552 | NP_001092140.1 | |
NM_001098671.1 | 2234 | Silent Mutation | GCC,GCT | A,A 552 | NP_001092141.1 | |
NM_001318398.1 | 2234 | Silent Mutation | GCC,GCT | A,A 407 | NP_001305327.1 | |
NM_153819.1 | 2234 | Silent Mutation | GCC,GCT | A,A 552 | NP_722541.1 | |
XM_005273707.3 | 2234 | Silent Mutation | GCC,GCT | A,A 657 | XP_005273764.3 | |
XM_011544718.1 | 2234 | Silent Mutation | GCC,GCT | A,A 657 | XP_011543020.1 | |
XM_011544720.1 | 2234 | Silent Mutation | GCC,GCT | A,A 552 | XP_011543022.1 | |
XM_011544721.1 | 2234 | Silent Mutation | GCC,GCT | A,A 552 | XP_011543023.1 | |
XM_011544722.1 | 2234 | Silent Mutation | GCC,GCT | A,A 552 | XP_011543024.1 | |
XM_011544723.2 | 2234 | Silent Mutation | GCC,GCT | A,A 552 | XP_011543025.1 | |
XM_011544725.2 | 2234 | Silent Mutation | GCC,GCT | A,A 407 | XP_011543027.1 | |
XM_017017082.1 | 2234 | Silent Mutation | GCC,GCT | A,A 840 | XP_016872571.1 | |
XM_017017083.1 | 2234 | Silent Mutation | GCC,GCT | A,A 840 | XP_016872572.1 | |
XM_017017084.1 | 2234 | Silent Mutation | GCC,GCT | A,A 552 | XP_016872573.1 | |
XM_017017085.1 | 2234 | Silent Mutation | GCC,GCT | A,A 493 | XP_016872574.1 | |
XM_017017086.1 | 2234 | Silent Mutation | GCC,GCT | A,A 407 | XP_016872575.1 |