Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGAGGGACTTTCTTTCCAAGTA[C/T]GGCATTCCCGATATACCAGAAAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SLC43A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC43A3 - solute carrier family 43 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278201.1 | 1774 | Missense Mutation | CAT,CGT | H,R 481 | NP_001265130.1 | |
NM_001278206.1 | 1774 | Missense Mutation | CAT,CGT | H,R 494 | NP_001265135.1 | |
NM_014096.3 | 1774 | Missense Mutation | CAT,CGT | H,R 481 | NP_054815.2 | |
NM_017611.2 | 1774 | Missense Mutation | CAT,CGT | H,R 481 | NP_060081.1 | |
NM_199329.2 | 1774 | Missense Mutation | CAT,CGT | H,R 481 | NP_955361.1 |