Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGAGGAACACGGCCACGGTGCC[C/G]GCCAGGAGCCTGGCTGCACTTCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600644 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NECTIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NECTIN1 - nectin cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002855.4 | 1225 | Intron | NP_002846.3 | |||
NM_203285.1 | 1225 | Silent Mutation | GCC,GCG | A,A 351 | NP_976030.1 | |
NM_203286.1 | 1225 | Intron | NP_976031.1 |