Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAATTTTATGGATACATTAAGCT[A/G]ATAAATTTTATTAGACTTAAGGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605628 MIM: 603190 | ||||||||||||||||||||
Literature Links: |
HTATIP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HTATIP2 - HIV-1 Tat interactive protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRMT3 - protein arginine methyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145166.1 | 493 | Intron | NP_001138638.1 | |||
NM_001145167.1 | 493 | Silent Mutation | CTA,CTG | L,L 15 | NP_001138639.1 | |
NM_005788.3 | 493 | Silent Mutation | CTA,CTG | L,L 92 | NP_005779.1 | |
XM_011519836.2 | 493 | Intron | XP_011518138.1 | |||
XM_017017081.1 | 493 | Intron | XP_016872570.1 |