Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCAGCTGCGAGCCCCCTCGCATT[C/T]GCGGAGCCGGGACACGAGGTGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118445 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCKBR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCKBR - cholecystokinin B receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318029.1 | 451 | Missense Mutation | CGC,TGC | R,C 45 | NP_001304958.1 | |
NM_176875.3 | 451 | Missense Mutation | CGC,TGC | R,C 45 | NP_795344.1 | |
XM_005253210.1 | 451 | Missense Mutation | CGC,TGC | R,C 45 | XP_005253267.1 | |
XM_017018516.1 | 451 | Intron | XP_016874005.1 |