Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602647 MIM: 602946 | ||||||||||||||||||||
Literature Links: |
MIR6514 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6514 - microRNA 6514 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6748 - microRNA 6748 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NXF1 - nuclear RNA export factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF6L - TATA-box binding protein associated factor 6 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM179B - transmembrane protein 179B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199337.2 | 675 | Missense Mutation | CGC,TGC | R,C 215 | NP_955369.1 | |
XM_005273982.3 | 675 | UTR 3 | XP_005274039.3 |
TMEM223 - transmembrane protein 223 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080501.2 | 675 | Intron | NP_001073970.1 |