Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTCTCAGGGTGCAGTGGTGCCC[A/G]GATGTCCAGTCTGCGAGTCTTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616254 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLPB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLPB - ClpB homolog, mitochondrial AAA ATPase chaperonin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258392.2 | 2293 | Missense Mutation | CGG,TGG | R,W 665 | NP_001245321.1 | |
NM_001258393.2 | 2293 | Missense Mutation | CGG,TGG | R,W 636 | NP_001245322.1 | |
NM_001258394.2 | 2293 | Missense Mutation | CGG,TGG | R,W 650 | NP_001245323.1 | |
NM_030813.5 | 2293 | Missense Mutation | CGG,TGG | R,W 695 | NP_110440.1 | |
XM_005274320.1 | 2293 | Missense Mutation | CGG,TGG | R,W 666 | XP_005274377.1 | |
XM_011545289.2 | 2293 | Intron | XP_011543591.1 |
LOC105369375 - uncharacterized LOC105369375 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |