Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCAGCCTGGGGGCCCGGCAGCA[A/G]CGACAGCAGCGGCCTGGAGGAGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118510 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRM1 - cholinergic receptor muscarinic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000738.2 | 885 | Silent Mutation | CGC,CGT | R,R 261 | NP_000729.2 | |
XM_011544742.2 | 885 | Silent Mutation | CGC,CGT | R,R 261 | XP_011543044.1 |