Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGTACCAGGAGACAAACCCATG[A/G]AGTGGGGTTGATGCTCACAGACTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603290 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHANK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SHANK2 - SH3 and multiple ankyrin repeat domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012309.4 | 7005 | UTR 3 | NP_036441.2 | |||
NM_133266.4 | 7005 | UTR 3 | NP_573573.2 | |||
XM_005277932.3 | 7005 | UTR 3 | XP_005277989.1 | |||
XM_017017387.1 | 7005 | UTR 3 | XP_016872876.1 | |||
XM_017017388.1 | 7005 | UTR 3 | XP_016872877.1 | |||
XM_017017389.1 | 7005 | UTR 3 | XP_016872878.1 | |||
XM_017017390.1 | 7005 | UTR 3 | XP_016872879.1 |