Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGCAGGACCGCCCACGCTTGAT[C/T]GGCGTGGGGTTGAGCACAGCCCGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615187 MIM: 179505 | ||||||||||||||||||||
Literature Links: |
PGAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PGAP2 - post-GPI attachment to proteins 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOG - ras homolog family member G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001665.3 | 881 | Silent Mutation | CCA,CCG | P,P 181 | NP_001656.2 | |
XM_005252916.2 | 881 | Silent Mutation | CCA,CCG | P,P 181 | XP_005252973.1 | |
XM_017017719.1 | 881 | Silent Mutation | CCA,CCG | P,P 181 | XP_016873208.1 |