Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTGCCCATTTAGTGTTTGGCTC[A/G]TTTGGTTCATGGGTCTCATTTGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607537 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAML2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAML2 - mastermind like transcriptional coactivator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032427.3 | 6215 | Missense Mutation | ACG,ATG | T,M 1032 | NP_115803.1 | |
XM_011543023.2 | 6215 | Missense Mutation | ACG,ATG | T,M 885 | XP_011541325.1 | |
XM_011543024.2 | 6215 | Missense Mutation | ACG,ATG | T,M 804 | XP_011541326.1 | |
XM_011543025.1 | 6215 | Intron | XP_011541327.1 |