Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTCACGGGTGATGGGAATGGTG[C/T]GCTCAGGGCCAGAGACCTGTTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123590 MIM: 602179 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYAB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRYAB - crystallin alpha B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289807.1 | 777 | Missense Mutation | CAC,CGC | H,R 157 | NP_001276736.1 | |
NM_001289808.1 | 777 | Missense Mutation | CAC,CGC | H,R 157 | NP_001276737.1 | |
NM_001885.2 | 777 | Missense Mutation | CAC,CGC | H,R 157 | NP_001876.1 | |
XM_011542608.1 | 777 | Missense Mutation | CAC,CGC | H,R 157 | XP_011540910.1 | |
XM_011542609.2 | 777 | Missense Mutation | CAC,CGC | H,R 90 | XP_011540911.1 |
HSPB2 - heat shock protein family B (small) member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSPB2-C11orf52 - HSPB2-C11orf52 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |