Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTGAGCAATGATGGGGCCGCTCC[C/T]GGGGCCACACCTGCTTCTGGATCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608549 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VPS11 - VPS11, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290185.1 | 117 | UTR 5 | NP_001277114.1 | |||
NM_021729.5 | 117 | Silent Mutation | CCC,CCT | P,P 29 | NP_068375.3 | |
XM_011542898.1 | 117 | Intron | XP_011541200.1 | |||
XM_011542899.2 | 117 | UTR 5 | XP_011541201.1 |