Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGTACGCAGGGCCTCAAAGAC[A/G]TCCCACCCGAGCCGACCCGAGACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602732 MIM: 616454 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP1 - Rho GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004308.3 | 123 | Intron | NP_004299.1 | |||
XM_011520095.1 | 123 | Intron | XP_011518397.1 | |||
XM_017017720.1 | 123 | Intron | XP_016873209.1 |
ZNF408 - zinc finger protein 408 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184751.1 | 123 | Missense Mutation | ATC,GTC | I,V 35 | NP_001171680.1 | |
NM_024741.2 | 123 | Missense Mutation | ATC,GTC | I,V 43 | NP_079017.1 |