Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTATTACCTCTGGGTCTCTGGA[A/G]TATGTCTTGCCCCTTTGACGAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615584 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM111B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM111B - family with sequence similarity 111 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142703.1 | Intron | NP_001136175.1 | ||||
NM_001142704.1 | Intron | NP_001136176.1 | ||||
NM_198947.3 | Intron | NP_945185.1 |