Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCTCATATGTCCTTTTACGTTT[C/T]TCCTCAATCCGGTGACGATCGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164014 MIM: 602180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4489 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4489 - microRNA 4489 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RELA - RELA proto-oncogene, NF-kB subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145138.1 | 808 | Silent Mutation | GAA,GAG | E,E 297 | NP_001138610.1 | |
NM_001243984.1 | 808 | Silent Mutation | GAA,GAG | E,E 300 | NP_001230913.1 | |
NM_001243985.1 | 808 | Silent Mutation | GAA,GAG | E,E 300 | NP_001230914.1 | |
NM_021975.3 | 808 | Silent Mutation | GAA,GAG | E,E 300 | NP_068810.3 | |
XM_011545206.1 | 808 | Silent Mutation | GAA,GAG | E,E 229 | XP_011543508.1 | |
XM_011545207.1 | 808 | Silent Mutation | GAA,GAG | E,E 194 | XP_011543509.1 |
SIPA1 - signal-induced proliferation-associated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |