Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAGTCAAGGGGCAGGTCAGCCC[C/T]GTAGGCCACGTAGGCTATGGCAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR2AT4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR2AT4 - olfactory receptor family 2 subfamily AT member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005285.1 | 2438 | Missense Mutation | AGG,GGG | R,G 268 | NP_001005285.1 | |
XM_017017655.1 | 2438 | Missense Mutation | AGG,GGG | R,G 268 | XP_016873144.1 | |
XM_017017656.1 | 2438 | Missense Mutation | AGG,GGG | R,G 268 | XP_016873145.1 | |
XM_017017657.1 | 2438 | Missense Mutation | AGG,GGG | R,G 268 | XP_016873146.1 | |
XM_017017658.1 | 2438 | Missense Mutation | AGG,GGG | R,G 268 | XP_016873147.1 |