Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTATGGGGGGCAGTTGGGATCATG[A/G]CTCGGGGTAGAGGTGAGACAGTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601783 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP6 - microtubule associated protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033063.1 | 5045 | Missense Mutation | GCC,GTC | A,V 798 | NP_149052.1 | |
NM_207577.1 | 5045 | Intron | NP_997460.1 | |||
XM_006718556.3 | 5045 | Intron | XP_006718619.1 | |||
XM_011545039.2 | 5045 | Intron | XP_011543341.1 | |||
XM_017017755.1 | 5045 | Missense Mutation | GCC,GTC | A,V 814 | XP_016873244.1 | |
XM_017017756.1 | 5045 | Missense Mutation | GCC,GTC | A,V 513 | XP_016873245.1 | |
XM_017017757.1 | 5045 | Missense Mutation | GCC,GTC | A,V 497 | XP_016873246.1 | |
XM_017017758.1 | 5045 | Missense Mutation | GCC,GTC | A,V 485 | XP_016873247.1 |