Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCGCGGCCATGGGGAGCTTGAA[A/G]GAGGAGCTGCTCAAAGCCATCTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604762 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SWAP70 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SWAP70 - SWAP switching B-cell complex 70kDa subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297714.1 | 118 | Silent Mutation | AAA,AAG | K,K 5 | NP_001284643.1 | |
NM_015055.3 | 118 | Silent Mutation | AAA,AAG | K,K 5 | NP_055870.2 | |
XM_005252830.4 | 118 | Intron | XP_005252887.1 |