Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTCCTGGGACCTTGCAGGATTT[C/T]GAACGCTGGACCGCGGACTCCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607104 | ||||||||||||||||||||
Literature Links: |
TNKS1BP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNKS1BP1 - tankyrase 1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033396.2 | 5226 | Silent Mutation | TCA,TCG | S,S 1687 | NP_203754.2 | |
XM_006718725.3 | 5226 | Silent Mutation | TCA,TCG | S,S 1687 | XP_006718788.1 | |
XM_011545325.2 | 5226 | Intron | XP_011543627.1 |