Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCAAACCCAGCAAGTATGTCCC[A/G]GTCTCTGCAGCCGCCATCTTCCTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614586 | ||||||||||||||||||||
Literature Links: |
ZDHHC5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZDHHC5 - zinc finger DHHC-type containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015457.2 | 1234 | Silent Mutation | CCA,CCG | P,P 16 | NP_056272.2 | |
XM_011544899.1 | 1234 | Silent Mutation | CCA,CCG | P,P 16 | XP_011543201.1 | |
XM_011544900.1 | 1234 | Silent Mutation | CCA,CCG | P,P 16 | XP_011543202.1 | |
XM_011544901.1 | 1234 | Silent Mutation | CCA,CCG | P,P 16 | XP_011543203.1 | |
XM_017017498.1 | 1234 | UTR 5 | XP_016872987.1 |