Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGCAGCTCTCGCCCGCGCAGC[A/G]GAAGTCGATCTCCTTGTGGTCGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602131 MIM: 602631 | ||||||||||||||||||||
Literature Links: |
PHLDA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHLDA2 - pleckstrin homology like domain family A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003311.3 | 303 | Missense Mutation | CGC,TGC | R,C 83 | NP_003302.1 |
SLC22A18 - solute carrier family 22 member 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |