Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCACCACAGCCCTACCGGGAGCC[A/G]GAACCCTCTGTGGCCGAACCCCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602313 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OVOL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OVOL1 - ovo like transcriptional repressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004561.3 | 481 | Silent Mutation | CCA,CCG | P,P 47 | NP_004552.2 | |
XM_005274018.3 | 481 | UTR 5 | XP_005274075.1 | |||
XM_011545067.2 | 481 | UTR 5 | XP_011543369.1 | |||
XM_011545068.2 | 481 | UTR 5 | XP_011543370.1 | |||
XM_017017837.1 | 481 | UTR 5 | XP_016873326.1 | |||
XM_017017838.1 | 481 | UTR 5 | XP_016873327.1 |
OVOL1-AS1 - OVOL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |