Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTACTACTGGTGGCTGAACACA[A/G]TGGTCTTCCCAGTCATGTATAACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609472 | ||||||||||||||||||||
Literature Links: |
CNGA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNGA4 - cyclic nucleotide gated channel alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037329.3 | 143 | Missense Mutation | ATG,GTG | M,V 41 | NP_001032406.1 | |
XM_017017217.1 | 143 | Missense Mutation | ATG,GTG | M,V 41 | XP_016872706.1 | |
XM_017017218.1 | 143 | Missense Mutation | ATG,GTG | M,V 1 | XP_016872707.1 | |
XM_017017219.1 | 143 | Missense Mutation | ATG,GTG | M,V 1 | XP_016872708.1 |
FAM160A2 - family with sequence similarity 160 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |