Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGACAAGTGCACCCCAGAACAC[A/G]ACCCAGCCCAAGCTCCTGGCTCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608403 | ||||||||||||||||||||
Literature Links: |
MS4A10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MS4A10 - membrane spanning 4-domains A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206893.3 | 204 | Silent Mutation | ACA,ACG | T,T 36 | NP_996776.2 | |
XM_011544989.1 | 204 | Silent Mutation | ACA,ACG | T,T 36 | XP_011543291.1 | |
XM_011544991.2 | 204 | Intron | XP_011543293.1 |