Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCCCAGTATGCAAAGTTCCAGC[A/G]GCTGCCGTCAACCCACTGAAAGCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605601 MIM: 606814 | ||||||||||||||||||||
Literature Links: |
PRG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRG2 - proteoglycan 2, pro eosinophil major basic protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243245.2 | 722 | Missense Mutation | CGC,TGC | R,C 168 | NP_001230174.1 | |
NM_001302926.1 | 722 | Missense Mutation | CGC,TGC | R,C 179 | NP_001289855.1 | |
NM_001302927.1 | 722 | Missense Mutation | CGC,TGC | R,C 179 | NP_001289856.1 | |
NM_002728.5 | 722 | Missense Mutation | CGC,TGC | R,C 179 | NP_002719.3 |
PRG3 - proteoglycan 3, pro eosinophil major basic protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |