Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGGTCAATGAGCTGAAGGAGAA[A/G]GGCAACAAGGCCCTGAGCGTGGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605063 | ||||||||||||||||||||
Literature Links: |
STIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STIP1 - stress induced phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282652.1 | 724 | Silent Mutation | AAA,AAG | K,K 57 | NP_001269581.1 | |
NM_001282653.1 | 724 | Intron | NP_001269582.1 | |||
NM_006819.2 | 724 | Intron | NP_006810.1 |