Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTTCTGCCGGCGGACGCAGCAG[C/G]AGGCGGCCAGCGTGGTGAGGAGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137207 | ||||||||||||||||||||
Literature Links: |
LRRC32 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC32 - leucine rich repeat containing 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128922.1 | 2275 | Missense Mutation | TCC,TGC | S,C 648 | NP_001122394.1 | |
NM_005512.2 | 2275 | Missense Mutation | TCC,TGC | S,C 648 | NP_005503.1 | |
XM_005273902.3 | 2275 | Missense Mutation | TCC,TGC | S,C 648 | XP_005273959.1 | |
XM_005273903.4 | 2275 | Missense Mutation | TCC,TGC | S,C 626 | XP_005273960.2 | |
XM_011544935.2 | 2275 | Missense Mutation | TCC,TGC | S,C 648 | XP_011543237.1 | |
XM_017017527.1 | 2275 | Missense Mutation | TCC,TGC | S,C 648 | XP_016873016.1 |