Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTCCAGAGTACAGGCTCTGATA[C/T]GTGACAGGCCTGTAGGTGGCCGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604648 | ||||||||||||||||||||
Literature Links: |
NUDT8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUDT8 - nudix hydrolase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBX10 - T-box 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005995.4 | 1356 | Silent Mutation | ACA,ACG | T,T 326 | NP_005986.2 | |
XM_011544992.2 | 1356 | Silent Mutation | ACA,ACG | T,T 371 | XP_011543294.1 | |
XM_011544993.1 | 1356 | Silent Mutation | ACA,ACG | T,T 165 | XP_011543295.1 |