Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATCTGGACTTTCATCAGGGCCC[A/G]TTTCAGCTGCTCATAGGTTACGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602044 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UCP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UCP3 - uncoupling protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003356.3 | 1250 | Missense Mutation | CGG,TGG | R,W 299 | NP_003347.1 | |
NM_022803.2 | 1250 | Intron | NP_073714.1 |