Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCGCCTGCTGTTACCTGGGGGA[A/C]TTCTTGGTGTGGCAGTGGTGGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602047 MIM: 602854 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE3B - phosphodiesterase 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000922.3 | 880 | Missense Mutation | GAA,GAC | E,D 165 | NP_000913.2 | |
XM_006718249.3 | 880 | Missense Mutation | GAA,GAC | E,D 165 | XP_006718312.1 | |
XM_017017911.1 | 880 | Missense Mutation | GAA,GAC | E,D 165 | XP_016873400.1 | |
XM_017017912.1 | 880 | Missense Mutation | GAA,GAC | E,D 165 | XP_016873401.1 |
PSMA1 - proteasome subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143937.1 | 880 | Intron | NP_001137409.1 | |||
NM_002786.3 | 880 | Intron | NP_002777.1 | |||
NM_148976.2 | 880 | Intron | NP_683877.1 |