Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAAAAGGTCATAGTTTAAGTATA[A/T]ACGAAGCAATTCAGCAGCATCAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607614 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP160 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUP160 - nucleoporin 160 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318399.1 | 4142 | Intron | NP_001305328.1 | |||
NM_015231.2 | 4142 | Missense Mutation | ATA,TTA | I,L 1340 | NP_056046.1 |