Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCTGGGAGAATCATGAGGCTG[G/T]GGACCTGGAGAGAGCGGCTGAGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614983 | ||||||||||||||||||||
Literature Links: |
BATF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BATF2 - basic leucine zipper ATF-like transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300807.1 | 551 | Silent Mutation | CCA,CCC | P,P 116 | NP_001287736.1 | |
NM_001300808.1 | 551 | Silent Mutation | CCA,CCC | P,P 55 | NP_001287737.1 | |
NM_138456.3 | 551 | Silent Mutation | CCA,CCC | P,P 140 | NP_612465.3 |