Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTTTCTGTCACAAAAGCTAGAA[A/G]AAAAGGTATAGTATGATTAGTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610315 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105369438 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105369438 - uncharacterized LOC105369438 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIWIL4 - piwi like RNA-mediated gene silencing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152431.2 | 719 | Missense Mutation | AAA,GAA | K,E 170 | NP_689644.2 |