Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCATTATTGCTTTGTTTTTATT[A/G]CTTGTAGGTAGTCCTATCTCTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PACS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PACS1 - phosphofurin acidic cluster sorting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018026.3 | 485 | Intron | NP_060496.2 | |||
XM_011545162.1 | 485 | Intron | XP_011543464.1 | |||
XM_011545164.2 | 485 | UTR 5 | XP_011543466.1 |