Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCACTTGAATTATCTTGTCGAA[C/T]ACGTTTGGCAAATCCAGAATTTACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611271 | ||||||||||||||||||||
Literature Links: |
KIF18A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF18A - kinesin family member 18A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031217.3 | 1450 | Missense Mutation | ATT,GTT | I,V 854 | NP_112494.3 | |
XM_017018379.1 | 1450 | Missense Mutation | ATT,GTT | I,V 854 | XP_016873868.1 | |
XM_017018380.1 | 1450 | Missense Mutation | ATT,GTT | I,V 410 | XP_016873869.1 |