Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCCACTAAGCTATAGTCCAATG[G/T]GCTCCCCACTTTGCCTACATTTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608522 | ||||||||||||||||||||
Literature Links: |
RSF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RSF1 - remodeling and spacing factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016578.3 | 3972 | Missense Mutation | ACA,CCA | T,P 1360 | NP_057662.3 | |
XM_005274051.2 | 3972 | Missense Mutation | ACA,CCA | T,P 1357 | XP_005274108.1 | |
XM_017017923.1 | 3972 | Missense Mutation | ACA,CCA | T,P 1319 | XP_016873412.1 |