Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTCGGGGAGCTGGCAATCGCGG[C/T]CGGAGGGCCCGGGCACCCAGGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602090 MIM: 607982 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LTBP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LTBP3 - latent transforming growth factor beta binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130144.2 | 3804 | Missense Mutation | GAC,GGC | D,G 1118 | NP_001123616.1 | |
NM_001164266.1 | 3804 | Intron | NP_001157738.1 | |||
NM_021070.4 | 3804 | Intron | NP_066548.2 | |||
XM_011545032.2 | 3804 | Missense Mutation | GAC,GGC | D,G 1127 | XP_011543334.1 | |
XM_011545033.2 | 3804 | Intron | XP_011543335.1 | |||
XM_017017737.1 | 3804 | Intron | XP_016873226.1 |
SCYL1 - SCY1 like pseudokinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |