Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACATCTCACCTCCCCTTAGCGG[A/G]GCAGAAACGTGAGTAGCGGGTGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615755 | ||||||||||||||||||||
Literature Links: |
FOXR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOXR1 - forkhead box R1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181721.2 | 278 | Missense Mutation | GAG,GGG | E,G 18 | NP_859072.1 | |
XM_017017575.1 | 278 | Missense Mutation | GAG,GGG | E,G 18 | XP_016873064.1 | |
XM_017017576.1 | 278 | Missense Mutation | GAG,GGG | E,G 18 | XP_016873065.1 |