Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCACTGGCGATGCTGGAGAAGAC[C/G]GTGGAAAAATGCTGCATCTCCCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176875 MIM: 603761 | ||||||||||||||||||||
Literature Links: |
LOC100130987 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100130987 - uncharacterized LOC100130987 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1CA - protein phosphatase 1 catalytic subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAD9A - RAD9 checkpoint clamp component A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243224.1 | 584 | Silent Mutation | ACC,ACG | T,T 17 | NP_001230153.1 | |
NM_004584.2 | 584 | Silent Mutation | ACC,ACG | T,T 93 | NP_004575.1 | |
XM_006718652.3 | 584 | Silent Mutation | ACC,ACG | T,T 104 | XP_006718715.1 | |
XM_017018097.1 | 584 | UTR 5 | XP_016873586.1 | |||
XM_017018098.1 | 584 | UTR 5 | XP_016873587.1 |