Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACTGGATGACCTGTTCCCCAAT[A/G]AGGATGAGCAGAGCCCAGGTGCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611729 MIM: 612565 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLC2 - kinesin light chain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134774.1 | 95 | Missense Mutation | AAG,GAG | K,E 94 | NP_001128246.1 | |
NM_001134775.1 | 95 | Missense Mutation | AAG,GAG | K,E 171 | NP_001128247.1 | |
NM_001134776.1 | 95 | Missense Mutation | AAG,GAG | K,E 171 | NP_001128248.1 | |
NM_001318734.1 | 95 | Missense Mutation | AAG,GAG | K,E 171 | NP_001305663.1 | |
NM_022822.2 | 95 | Missense Mutation | AAG,GAG | K,E 171 | NP_073733.1 | |
XM_005274183.1 | 95 | Missense Mutation | AAG,GAG | K,E 171 | XP_005274240.1 | |
XM_005274184.1 | 95 | Missense Mutation | AAG,GAG | K,E 171 | XP_005274241.1 | |
XM_017018133.1 | 95 | UTR 5 | XP_016873622.1 |
RAB1B - RAB1B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |