Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCATTGGCTGACTGTTCATGTG[G/T]GTCTGCATATGTGGGGGGGTGTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606985 MIM: 607108 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELP4 - elongator acetyltransferase complex subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAX6 - paired box 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000280.4 | 1455 | Silent Mutation | NP_000271.1 | |||
NM_001127612.1 | 1455 | Silent Mutation | NP_001121084.1 | |||
NM_001258462.1 | 1455 | Silent Mutation | NP_001245391.1 | |||
NM_001258463.1 | 1455 | Silent Mutation | NP_001245392.1 | |||
NM_001258464.1 | 1455 | Silent Mutation | NP_001245393.1 | |||
NM_001258465.1 | 1455 | Silent Mutation | NP_001245394.1 | |||
NM_001310158.1 | 1455 | Silent Mutation | NP_001297087.1 | |||
NM_001310159.1 | 1455 | Intron | NP_001297088.1 | |||
NM_001310160.1 | 1455 | Silent Mutation | NP_001297089.1 | |||
NM_001310161.1 | 1455 | Silent Mutation | NP_001297090.1 | |||
NM_001604.5 | 1455 | Silent Mutation | NP_001595.2 |