Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTATGGGGACTGTTGAGGATAA[C/T]CAGGACTGGGGGCCCTGCCTCCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153340 MIM: 610038 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD5 - CD5 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS37C - VPS37C, ESCRT-I subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017966.4 | 985 | Missense Mutation | GAT,GGT | D,G 295 | NP_060436.4 | |
XM_005274077.3 | 985 | Missense Mutation | GAT,GGT | D,G 295 | XP_005274134.1 | |
XM_017017955.1 | 985 | Missense Mutation | GAT,GGT | D,G 182 | XP_016873444.1 |