Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCACTTGAACATAGGATTTTTGC[A/G]CTGTGCTTCCTCTAGGACAACAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609342 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GIF - gastric intrinsic factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005142.2 | 1088 | Missense Mutation | CGC,TGC | R,C 352 | NP_005133.2 | |
XM_011544939.2 | 1088 | Missense Mutation | CGC,TGC | R,C 338 | XP_011543241.1 |