Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAAAGTTGCGGGCCAGCACACA[C/G]ACCAGCACATTGCCCAAGAGCTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 141900 MIM: 142000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HBB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HBB - hemoglobin subunit beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HBBP1 - hemoglobin subunit beta pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HBD - hemoglobin subunit delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000519.3 | 531 | Silent Mutation | GTC,GTG | V,V 112 | NP_000510.1 |