Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTCCCCCAGCTGCACCCTGGG[A/C]CTGGTCTTGCCCCTCTGGAGTGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615126 MIM: 606780 | ||||||||||||||||||||
Literature Links: |
ANKRD13D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD13D - ankyrin repeat domain 13D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSH3 - slingshot protein phosphatase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017857.3 | 664 | Silent Mutation | GGA,GGC | G,G 162 | NP_060327.3 | |
XM_017017942.1 | 664 | Silent Mutation | GGA,GGC | G,G 62 | XP_016873431.1 |