Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTGGGGAAGTTAGCTGTGAAGA[C/T]AATTGTTCCTCCTGCATCATCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAML PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JAML - junction adhesion molecule like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098526.1 | Intron | NP_001091996.1 | ||||
NM_001286570.1 | Intron | NP_001273499.1 | ||||
NM_001286571.1 | Intron | NP_001273500.1 | ||||
NM_153206.2 | Intron | NP_694938.2 | ||||
XM_005271407.3 | Intron | XP_005271464.1 | ||||
XM_017017213.1 | Intron | XP_016872702.1 | ||||
XM_017017214.1 | Intron | XP_016872703.1 | ||||
XM_017017215.1 | Intron | XP_016872704.1 |