Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTGTTTTTCTCAAACTGTGTGT[A/G]GTCTGGGTAAGCCTCTTTCACGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604773 MIM: 613739 MIM: 610027 | ||||||||||||||||||||
Literature Links: |
ACAD8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACAD8 - acyl-CoA dehydrogenase family member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THYN1 - thymocyte nuclear protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037304.1 | 511 | Missense Mutation | CAC,TAC | H,Y 137 | NP_001032381.1 | |
NM_001037305.1 | 511 | Missense Mutation | CAC,TAC | H,Y 137 | NP_001032382.1 | |
NM_014174.2 | 511 | Missense Mutation | CAC,TAC | H,Y 137 | NP_054893.1 | |
NM_199297.1 | 511 | Missense Mutation | CAC,TAC | H,Y 137 | NP_954994.1 | |
NM_199298.1 | 511 | Missense Mutation | CAC,TAC | H,Y 137 | NP_954995.1 | |
XM_005271527.3 | 511 | Missense Mutation | CAC,TAC | H,Y 178 | XP_005271584.1 | |
XM_005271528.3 | 511 | Missense Mutation | CAC,TAC | H,Y 178 | XP_005271585.1 |
VPS26B - VPS26, retromer complex component B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |