Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAACTTGGTGAAGGTGTTGATG[A/G]GATTAATGGCCTTGAGGGTGATGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602732 MIM: 615088 | ||||||||||||||||||||
Literature Links: |
ARHGAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP1 - Rho GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004308.3 | 1161 | Missense Mutation | CCC,CTC | P,L 415 | NP_004299.1 | |
XM_011520095.1 | 1161 | Missense Mutation | CCC,CTC | P,L 415 | XP_011518397.1 | |
XM_017017720.1 | 1161 | Missense Mutation | CCC,CTC | P,L 371 | XP_016873209.1 |
ATG13 - autophagy related 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |