Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTAAAATCCATTTCACTAACCA[C/T]GTGCTTCCACATCATATTCTTCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602158 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLNS1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLNS1A - chloride nucleotide-sensitive channel 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293.2 | 562 | Intron | NP_001284.1 | |||
NM_001311199.1 | 562 | Missense Mutation | CAT,CGT | H,R 157 | NP_001298128.1 | |
NM_001311200.1 | 562 | Intron | NP_001298129.1 | |||
NM_001311201.1 | 562 | Intron | NP_001298130.1 | |||
NM_001311202.1 | 562 | Intron | NP_001298131.1 |
LOC107987163 - uncharacterized LOC107987163 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |